Human Rim3 Peptide (17 aa near N-terminus)
Catalog # 89419-636
Supplier: Prosci
CAS Number:
Specifications
- Species:Human
- Size:0.05 mg
- Storage Conditions:Store at –20 °C, stable for one year.
- Amino Acid Number:17 aa near N-terminus
- Protein/Peptide Name:Rim3 Peptide (17 aa near N-terminus)
- Tested Applications:Blocking
- Cat. No.:89419-636
- Supplier no.:4471P
Specifications
About this item
Rim3 peptide is used for blocking the activity of Slitrk6 antibody.