ContactUsLinkComponent
ImmEdge™ Hydrophobic Barrier Pen, Vector Laboratories
  101098-065
 :  
undefined
ImmEdge™ Hydrophobic Barrier Pen, Vector Laboratories
  101098-065
 :  H-4000
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    ImmEdge™ Hydrophobic Barrier Pen
  • Size:
    2-pen set (Each)
  • Cat. No.:
    101098-065
  • Supplier no.:
    H-4000
The ImmEdge™ Hydrophobic Barrier Pen can be used to draw a barrier around a section on a slide that keeps aqueous solutions in place and minimizes the amount of reagent needed for each section.
  • Heat stable
  • Stable for use with buffers with and without detergent (Tween 20, Triton X-100, etc.)
  • Keeps reagents localized on cells and tissue preparations
  • Light blue barrier
  • Compatible with enzyme or fluorescence-based detection systems
  • Soluble in all commonly used clearing agents
  • Environmentally friendly - does not contain "ozone-depleting" substances
The ImmEdge™ Pen is an improvement over the PAP pen for immunohistochemistry, immunocytochemistry, immunofluorescence, and in situ hybridization. It is designed to provide a heat-stable, water-repellent barrier that keeps reagents localized on tissue specimens and prevents mixing of reagents when multiple sections are mounted on the same slide. The ImmEdge™ Pen barrier is stable for use with buffers with and without detergent (e.g. Tween 20 or Triton X-100, etc.).
The pale blue barrier is insoluble in alcohol and acetone but is completely removed by all commonly used xylene and xylene substitute clearing agents. The ImmEdge™ Pen is compatible with enzyme or fluorescence-based detection systems.
The ImmEdge™ Pen is environmentally friendly. It does not contain "ozone-depleting" solvents as identified by the Montreal Protocol or the Council of the European Union (specifically, fluorocarbons and chlorocarbons).