ContactUsLinkComponent
Pierce™ Fluorescence Biotin Quantitation Kit, Thermo Scientific
Pierce™ Fluorescence Biotin Quantitation Kit, Thermo Scientific
Catalog # PI46610
Supplier:  Invitrogen
CAS Number:  
undefined
Pierce™ Fluorescence Biotin Quantitation Kit, Thermo Scientific
Catalog # PI46610
Supplier:  Invitrogen
Supplier Number:  46610
CAS Number:  
Specifications
  • Description:
    Fluorescence Biotin Quantitation Kit, sufficient for testing biotinylation of up to 50 protein samples tested all at once, or for approx. 5 separate uses
  • Size:
    200 Microplate Assays
  • Cat. No.:
    PI46610
  • Supplier no.:
    46610
Specifications
About this item
Thermo Scientific™ Pierce Fluorescence Biotin Quantitation Kit requires only ten microliters of sample to accurately measure the biotinylation level of labeled antibodies and other biomolecules.
  • Fast – requires only 5 minute incubation
  • Economical – requires only 10 µl of sample (~ 750 ng of biotinylated IgG)
  • Sensitive – linear working range of 10 to 60 picomoles of biotin
This microplate-based biotin assay is easy to perform by adding the supplied fluorescent reporter to the biotinylated samples and diluted biocytin standards. The avidin fluoresces when the weakly interacting HABA (4'-hydroxyazobenzene-2-carboxylic acid) is displaced by the biotin. The amount of biotin is determined by comparing the sample's fluorescence to the biocytin standard curve. This assay requires much less sample volume than the microplate colorimetric HABA assay and is much more sensitive.