ContactUsLinkComponent
Cleanline® Sticky Mat, Controlled Environment, Scapa Tapes NA LLC.
  33500-316
 :  Avantor
Cleanline® Sticky Mat, Controlled Environment, Scapa Tapes NA LLC.
  33500-316
 :  Avantor
  • Description:
    Cleanline® Sticky Mat
  • Length:
    114 cm (45")
  • Cat. No.:
    33500-316
  • Width:
    65 cm (25.5")
  • Supplier no.:
    CSD254546FBB
For over thirty years, Cleanline® Sticky Mats have been the standard for cleaning shoe soles before entry into cleanrooms.
  • Polyethylene film with aspecially treated pressure sensitive adhesive on one side, upon which persons entering a cleanroom or clean zone walk (and vehicles are rolled) to remove the last trace of contamination on shoe heels and soles, and on wheels
  • When a sheet becomes loaded with contamination, it is peeled off, exposing a new clean sheet for use, thus eliminating messy and time-consuming cleaning and washing
They have proven effective in a wide variety of industries, including microelectronics, aerospace, pharmaceutical, military, food, hospitals, and many others.