FuGENE® 6 Transfection Reagent, Promega
Supplier: Promega Corporation
Certificates
About this item
FuGENE® 6 Transfection Reagent is a nonliposomal formulation designed to transfect plasmid DNA into a wide variety of cell lines with high efficiency and low toxicity.
- Reliable Transfection for Common Cell Lines
- Preserve cell viability
- Excellent compatibility with everyday cell lines
- Straightforward protocol; minimal optimization
FuGENE® 6 Transfection Reagent is a nonliposomal formulation designed to transfect plasmid DNA into a wide variety of cell lines with high efficiency and low toxicity. The protocol does not require removal of serum or culture medium and does not require washing or changing of medium after introducing the reagent/DNA complex.
FuGENE is a registered trademark of Fugent, LLC, USA.
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together