Super Adjustable 2™ Super Erecta Shelf® Wire Shelving, Metroseal 3™ Finish, Metro
Supplier: Metro
Certificates
About this item
Metroseal 3 Corrosion-resistant shelves are electroplated carbon steel, with a special epoxy coating.
- Metroseal 3™ shelves are an economical alternative to stainless steel
- Metroseal 3™ shelves includes antimicrobial product protection, which inhibits the growth of bacteria, mold, mildew, and fungi that cause odors, stains, and product degradation
- Use within coolers to resist corrosion from moisture
- Shelves are not cart-washable
Open-wire design minimizes dust accumulation, allows a free circulation of air, and provides greater visibility of stored items. Shelves up to 121.9 cm (48") in length can support up to 363 kg (800 lbs.) per level. Shelves longer than 121.9 cm (48") and all super wide shelves can support up to 272 kg (600 lbs.) per level.
Certifications: Shelves meet MIL-S-40144E specifications.
Ordering information: Each shelf and post is sold separately. To order posts or accessories, contact your Avantor representative for more information. Each shelf includes four pairs of plastic split sleeves for securing shelf to posts. Manufacturer's 12-year limited warranty against rust formation.
Posts (Sold Individually)
Product Information
Frequently Bought Together