Butterfield's Phosphate Buffer, Hardy Diagnostics
89498-946
Product available on GSA Advantage®
:
- Pack type:Bottle
- Description:Ph 7.2
- Size:500ml
- Storage Temperature:2-8C
- Minimum Order Quantity:10
- Cat. No.:89498-946
- Fill volume:225 ml
- Supplier no.:U123
Butterfield's Phosphate Buffer, 225ml fill, 500ml polycarbonate wide mouth bottle with cap, order by the package of 10, by Hardy Diagnostics. MINIMUM ORDER: 3 packs.
(Butterfield's Phosphate Buffer, pH: 7.2) For the preparation of dilutions for plate count and other laboratory tests.
Butterfield's Phosphate Buffer, 225ml fill, 500ml polycarbonate wide mouth bottle with cap, order by the package of 10, by Hardy Diagnostics
MINIMUM ORDER: 3 packs.