Order Entry
United States
ContactUsLinkComponent
Letheen Broth, Modified, Hardy Diagnostics
Letheen Broth, Modified, Hardy Diagnostics
Catalog # 89497-620
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
Letheen Broth, Modified, Hardy Diagnostics
Catalog # 89497-620
Supplier:  Hardy Diagnostics
Supplier Number:  K293
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Pack type:
    Tube
  • Description:
    Letheen Broth, Modified
  • Size:
    16x125mm
  • Storage Temperature:
    2-8C
  • Minimum Order Quantity:
    5
  • Cat. No.:
    89497-620
  • Fill volume:
    9 ml
  • Application:
    For determination of antimicrobial activity of quaternary ammonium compounds and in the microbial evaluation of cosmetics.
  • Supplier no.:
    K293

Specifications

About this item

Letheen Broth, Modified, 16x125mm tube, 9ml fill, order by the package of 20, by Hardy Diagnostics.

Used to determine the antimicrobial activity of quaternary ammonium compounds and in the microbial evaluation of cosmetics.