Order Entry
United States
ContactUsLinkComponent
BCYE Agar without Cysteine, Hardy Diagnostics
BCYE Agar without Cysteine, Hardy Diagnostics
Catalog # 89498-574
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
BCYE Agar without Cysteine, Hardy Diagnostics
Catalog # 89498-574
Supplier:  Hardy Diagnostics
Supplier Number:  G108
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Pack type:
    Plate
  • Description:
    BCYE Agar without Cysteine
  • Size:
    15x100mm
  • Storage Temperature:
    2-8C
  • Minimum Order Quantity:
    10
  • Cat. No.:
    89498-574
  • Fill volume:
    18 ml
  • Application:
    For the cultivation of Legionella spp. from clinical and environmental specimens.
  • Supplier no.:
    G108

Specifications

About this item

(Buffered Charcoal Yeast Extract Agar) For the cultivation of Legionella spp. from clinical and environmental specimens.

(Buffered Charcoal Yeast Extract Agar) For the cultivation of Legionella spp

Conforms to ISO11371 formulation

from clinical and environmental specimens.

Ordering information: BCYE Agar without Cysteine, 15x100mm plate, order by the package of 10, by Hardy Diagnostics. Minimum Order: 5 packs