Specifications
- Chemical method:Salicylate
- Environmentally Preferable:
- Method:Color Disc/Salicylate, CD Platform
- Cat. No.:89500-732
- Range:0.1 - 2.5 mg/L NH₃-N
- Supplier no.:2428700
- Case:D
- No. of tests:100
Specifications
About this item
Designed for testing ammonia in freshwater or seawater, this kit's color disc method offers low-cost, fast, reliable results in an easy-to-carry kit.
Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.