ContactUsLinkComponent
Model PO-24 Phosphate Test Kit, Total Ortho-/Meta-, Hach
Model PO-24 Phosphate Test Kit, Total Ortho-/Meta-, Hach
  89500-712
 :  Avantor
Product available on GSA Advantage®
 :  
undefined
Model PO-24 Phosphate Test Kit, Total Ortho-/Meta-, Hach
  89500-712
 :  Avantor
Product available on GSA Advantage®
 :  
  • Chemical method:
    Ascorbic acid
  • Method:
    Color disc / Ascorbic Acid
  • Cat. No.:
    89500-712
  • Range:
    0.1 - 5 and 1 - 40
  • Supplier no.:
    225001
  • No. of tests:
    50 clear, 50 turbid
Kit provides measurement of ortho- (reactive) and meta- (condensed) phosphate (requires hot acid digestion).
Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.
 : Kit includes five reagents, color disc, comparator box, viewing adapter, two viewing tubes, bottle, flask, dropper tube, stove and cover, test tube clamp, funnel, filter paper, deionized water, instructions and carrying case.