Specifications
- Chemical method:Thymol blue
- Environmentally Preferable:
- Method:Drop Count Titration/ Thymolphthalein
- Cat. No.:89500-686
- Supplier no.:2183100
- No. of tests:100
Specifications
About this item
Formaldehyde solutions are used for maintenance of reverse osmosis systems and as disinfectants
- Utilizes drop count titration - a simple, economical titration method
- Kit contains all required reagents and apparatus in a rugged carrying case for analysis in the field
Hazardous Items. Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.
The Model FM-1 measures higher concentrations by drop count titration. Contains 100 tests.