Square Petri Dish with Grid, Simport
Supplier: Simport Scientific
Certificates
About this item
These square petri dishes are free from optical distortion.
They are ideal for phage typing, susceptibility testing, plate counts, and probe assats. Dishes feature an alphanumeric grid with four venting ribs on the underside of the lid to prevent build-up of condensation. Dishes are available sterile or nonsterile.
Frequently Bought Together