Order Entry
United States
ContactUsLinkComponent
Monoject™ Curved Tip Syringe, Covidien
Monoject™ Curved Tip Syringe, Covidien
Catalog # MJ8881-412012
Supplier:  Cardinal Health
Monoject™ Curved Tip Syringe, Covidien
Catalog # MJ8881-412012
Supplier:  Cardinal Health
Supplier Number:  8881412012

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Volume:
    12 ml
  • Description:
    Curved Tip Syringe
  • Cat. No.:
    MJ8881-412012
  • Tip style:
    Curved
  • Supplier no.:
    8881412012

Specifications

About this item

This Monoject™ specialty syringe is ungraduated and features a 12mL capacity

Equipped with a reusable plastic finger grip extender, it is ideal for pressure irrigation.