Ammonium cerium(IV) nitrate standard solution 0.5N, cartridge for digital titrator
97026-234
: Avantor
Product available on GSA Advantage®
:
- Pack type:Cartridge
- Size:1 X
- Environmentally Preferable:
- Cat. No.:97026-234
- Supplier no.:2270701
- Description:Ceric Standard Solution Titration Cartridge, 0.5 N
- Method:8351
- Method Name:Ceric Acid Titration
- Number of tests:25
- Parameter:Nitrite
- Platform:Digital Titrator
- Range:100 - 2500 mg/L NaNO2
- Titrant:0.5 N Ceric Ion
: Ammonium ceric nitrate, Ammonium hexanitratocerate (IV), Ceric Ammonium Nitrate, Cerium (IV) ammonium nitrate, Diammonium hexanitratocerate
:
:548.22 g/mol
:3264
:GHS03
:GHS05
:GHS07
:16774-21-3
:(NH₄)₂Ce(NO₃)₆
:MFCD00151121
:8 III
:13,00509