
Sodium hydroxide 8.00 N, cartridge for digital titrator
Supplier: Hach
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Storage Temperature:Ambient
UN:1824
ADR:8,II
:GHS05
Specification Test Results
- Method:8200 (Acid)
- Method Name:Phenolphthalein End Point
- Method Number 2:8233 (Base)
- Number of tests:40 - 100
- Parameter:Acid-Base
- Platform :Digital Titrator
- Range:10 - 4000 meq/L Acid
- Titrant:8 N NaOH
Specification Test Results
Frequently Bought Together