Sodium thiosulfate 0.200 N, cartridge for digital titrator
Supplier: Hach
Certificates
About this item
CAS Number:7772-98-7
:
Formula:Na₂S₂O₃
MDL Number:MFCD00003499
:158.11 g/mol
Storage Temperature:Ambient
Specification Test Results
- Method:8215
- Method Name:Azide mod. of Winkler Method
- Number of tests:40 - 100
- Parameter:Oxygen, Dissolved
- Platform :Digital Titrator
- Range:1 - 10 mg/L DO
- Titrant:0.200 N Na2S2O3
Specification Test Results
Frequently Bought Together