ContactUsLinkComponent
CitriSolv™ Xylene Substitute, for Histology and Pathology
CitriSolv™ Xylene Substitute, for Histology and Pathology
Catalog # 89426-268
Supplier:  Decon Labs
CAS Number:  
undefined
CitriSolv™ Xylene Substitute, for Histology and Pathology
Catalog # 89426-268
Supplier:  Decon Labs
Supplier Number:  1601
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Xylene substitute, CitriSolv™
  • Size:
    1 gal.
  • Cat. No.:
    89426-268
  • Supplier no.:
    1601
Specifications
About this item
CitriSolv™ is a d-Limonene-based solvent and clearing agent that can be used as a safe alternative to xylene and ethyl acetate
  • Safe alternative to xylene and ethyl acetate
  • Will not harden or shrink tissue
  • Biodegradable and non-corrosive
  • Does not contain benzene and has low levels of toxicity
It is soluble with absolute alcohol and will dissolve paraffin, glues, adhesives and mounting media. CitriSolv™ provides for better tissue sections - it will not harden or shrink tissue. It dries slowly with no residue and enhances some stains. Use for manual or automatic tissue processor staining procedures in histology/cytology applications that require xylene.