MICROFLEX® NeoTouch™ 25-201, Chemical-Resistant Gloves, Neoprene, Ansell
This glove is made with neoprene, an alternative to natural rubber latex that protects against Type I allergies.
- Powder-free
- Excellent chemical resistance
- Superior comfort
- Roll-beaded cuffs
These gloves provide excellent resistance to acids, bases and alcohols.
Ideal applications include assembly and inspection, lab analysis and technical work, and pharmaceutical manufacturing - especially when resistance to acids is required.
: This is an exam- and food-grade glove and complies with FDA food handling requirments (21 CFR 177.2600).
- Catalog No:
- 89412-174
- 89412-172
- 89412-170
- 89501-594
- 89501-596
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable/Reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
Frequently Bought Together