Microcentrifuge Septum Screw Caps Closure for Micrewtubes™ Centrifuge Tubes, Simport
89499-234
- Color:Aqua
- Description:Septum Screw Cap
- Cat. No.:89499-234
- Supplier no.:T347AQX
The screw cap incorporates a pierceable septum made of chemically resistant PTFE on the outside and silicone on the inside, both components being stable over a broad range of temperatures
The septum also acts as a silicone O-Ring for better sample protection. It is especially made to fit and be used with all Simport Micrewtubes™ Microcentrifuge Tubes. The cap is pierceable with pipet tips as well as with syringe needles.