ContactUsLinkComponent
OPTI-LYNX™ In-Line Trap / Filter Holder Kit
OPTI-LYNX™ In-Line Trap / Filter Holder Kit
Catalog # 83008-166
Supplier:  Optimize
undefined
OPTI-LYNX™ In-Line Trap / Filter Holder Kit
Catalog # 83008-166
Supplier:  Optimize
Supplier Number:  11-02225-AA
Specifications
  • Description:
    OPTI-LYNX™ In-line trap/filter holder kit (OPTI-LYNX™ filter/traps cartridges sold separately)
  • Environmentally Preferable:
  • Cat. No.:
    83008-166
  • Supplier no.:
    11-02225-AA
Specifications
About this item
The OPTI-LYNX™ In-Line Holder can be connected to ¹/₁₆" tubing for in-line filtration or trapping anywhere on your HPLC system.
  • For use with OPTI-LYNX™ Traps and Filters
  • Quarter-turn quick-connect design rated to 6000 psi
  • Low Volume/ Low Dispersion interface for volume critical regions
  • True zero-dead-volume connection
  • Hardware is only plumbed once
  • Hand-tight - no tools required
The OPTI-LYNX™ trapping configuration can also be placed directly within the loop of an injection valve to facilitate automated loading, flushing and elution. With Opti-Lynx, accessing and changing the insert takes only a quarter turn. Replace traps, filters or change configurations in seconds - without tools. OPTI-LYNX™ traps and filters (ordered separately) come in several bed volume/ diameter/ porosity combinations, allowing you to find your own balance between capacity and volume.
Delivery: Includes bayonet connector, holder cap, holder tube and fitting.