



Boron hydride-tetrahydrofuran complex 1 mol/l in THF stabilized for synthesis, Sigma-Aldrich®
Catalog # EM8.41077.0100
Supplier: MilliporeSigma
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Size:100 ml
- Cat. No.:EM8.41077.0100
- Supplier no.:8.41077.0100
Specifications
Specification Test Results
- Boiling Point:66 °C (1013 hPa) (THF)
- Flash Point:-21.7 °C
- Explosion Limit:1.58 - 12.42 %(V)
- Density:0.876 g/cm3 (25 °C)
- Ignition Point:103 °C
- Hazard Class:4.3
- Hazard Class Description:Dangerous When Wet Material
Specification Test Results
About this item
:
Boiling Pt:66 °C (1013 hPa) (THF)
Density:0.876 g/cm³ (25 °C)
EINECS:243-506-9
ADR:4.3,I
:M-Clarity™ quality level = MQ200
Flash Pt:–21.7 °C
UN:3148
:GHS02
:GHS05
:GHS08
:GHS07
Storage Temperature:Refrigerator