Order Entry
United States
ContactUsLinkComponent
Polyethylene glycol 6,000 for synthesis, Sigma-Aldrich®
Polyethylene glycol 6,000 for synthesis, Sigma-Aldrich®
Supplier:  MilliporeSigma
Certificates
Polyethylene glycol 6,000 for synthesis, Sigma-Aldrich®
Supplier:  MilliporeSigma
Certificates

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specification Test Results

  • Solubility:
    500 g/l (20 °C)
  • Melting Point:
    55 - 60 °C
  • Molar Mass:
    8000 g/mol
  • Vapor Pressure:
     - 0.01 hPa (20 °C)
  • Flash Point:
    250 °C
  • pH:
    5 - 7 (100 g/l, H2O, 20 °C)
  • Ignition Point:
    420 °C
  • Viscosity kinematic:
    200 - 250 mm2/s (20 °C) (50% solution)

Specification Test Results

Related Information

Frequently Bought Together