ContactUsLinkComponent
Multi-Element Standard VII (Cationic) for Ion Chrmatography (IC), Certipur®, MilliporeSigma
Multi-Element Standard VII (Cationic) for Ion Chrmatography (IC), Certipur®, MilliporeSigma
  EM1.10322.0100
 :  
Multi-Element Standard VII (Cationic) for Ion Chrmatography (IC), Certipur®, MilliporeSigma
  EM1.10322.0100
 :  1.10322.0100
 :  
  • Pack type:
    Plastic Bottle
  • Size:
    100 ml
  • Cat. No.:
    EM1.10322.0100
  • Supplier no.:
    1.10322.0100
9 cations: 100 mg/l: Li, Na, K, NH4, Mn, Ca, Mg, Sr, Ba