Order Entry
United States
ContactUsLinkComponent
Magnesium Standard, Titrisol®, MilliporeSigma
Magnesium Standard, Titrisol®, MilliporeSigma
Catalog # EM1.09949.0001
Supplier:  MilliporeSigma
CAS Number:  
Magnesium Standard, Titrisol®, MilliporeSigma
Catalog # EM1.09949.0001
Supplier:  MilliporeSigma
Supplier Number:  1.09949.0001
CAS Number:  

Specifications

  • Concentration:
    1000 mg
  • Matrix:
    6% HCl
  • Pack type:
    Plastic Ampule
  • Description:
    Concentrate
  • Size:
    1 Ampule
  • Cat. No.:
    EM1.09949.0001
  • Element:
    Magnesium
  • Supplier no.:
    1.09949.0001

Specifications

About this item