Order Entry
United States
ContactUsLinkComponent
Oxalic acid concentrate for dilution to 0.005 mol/L (0.01 N), Titrisol®, Supelco®
Oxalic acid concentrate for dilution to 0.005 mol/L (0.01 N), Titrisol®, Supelco®
Catalog # EM1.09932.0001
Supplier:  MilliporeSigma
CAS Number:  
Oxalic acid concentrate for dilution to 0.005 mol/L (0.01 N), Titrisol®, Supelco®
Catalog # EM1.09932.0001
Supplier:  MilliporeSigma
Supplier Number:  1.09932.0001
CAS Number:  

Specifications

  • Pack type:
    Cardboard box
  • Size:
    1 ampule
  • Cat. No.:
    EM1.09932.0001
  • Supplier no.:
    1.09932.0001

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Density:
    1.002 g/cm3 (20 °C)
  • pH:
    1.5 (H2O, 20 °C)

Specification Test Results

About this item

 :
 :90.04 g/mol
Storage Temperature:Ambient
ADR:8,III
 :Dose for preparation of 1 litre of solution.
EINECS:205-634-3
CAS Number:144-62-7
Formula:C₂H₂O₄
MDL Number:MFCD00002573