

Hanus solution (Iodine bromine) c(IBr) = 0.1 mol/l, Titripur® for determination of iodine value, Supelco®
Catalog # EM1.09164.1000
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:1 L
- Cat. No.:EM1.09164.1000
- Supplier no.:1.09164.1000
Specifications
Specification Test Results
- Form:liquid
- Amount-of-substance concentration:0.0995 - 0.1005
- Measurement uncertainty:+/- 0.0003
- Traceability:NIST SRM
- The concentration is determined by volumetric titration and refers to 20°C.:Corrosive Material
Specification Test Results
About this item
:
Density:1.06 g/cm3 (20 °C)
Flash Pt:40 °C
ADR:8 (3),II
:M-Clarity™ quality level = MQ100
UN:2789
:GHS02
:GHS05
Storage Temperature:Ambient
:7789-33-5
Formula:IBr
MDL Number:MFCD00011353