

Potassium hydroxide 0.1 mol/l (0.1 N) in isopropanol (IPA), Titripur® volumetric solution, Supelco®
EM1.05544.1000
:
- Pack type:Glass bottle
- Size:1 L
- Cat. No.:EM1.05544.1000
- Supplier no.:1.05544.1000
- Solubility:(20 °C) soluble
- Flash Point:12 °C (2-Propanol)
- Explosion Limit:2 - 13.4 %(V) (2-Propanol)
- Density:0.79 g/cm3 (20 °C)
- pH:13 (H2O, 20 °C)
- Ignition Point:425 °C (2-Propanol)
- Hazard Class:3
- Hazard Class Description:Flammable Liquid
:
:56.11 g/mol
:3 (8),II
:M-Clarity™ quality level = MQ100
:215-181-3
:2924
:GHS02
:GHS07
:Ambient
:1310-58-3
:KOH
:MFCD00003553