
Zinc iodide and starch solution analytical reagent, Supelco®
EM1.05445.0500
:
- Pack type:Plastic bottle
- Size:500 ml
- Cat. No.:EM1.05445.0500
- Supplier no.:1.05445.0500
- Solubility:(20 °C) soluble
- Density:1.02 g/cm3 (20 °C)
- pH:6.2 (H2O, 20 °C)
:
:GHS07
:MFCD00148838