

Iron(III) chloride hexahydrate, EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
Certificates
About this item
CAS Number:10025-77-1
:
Formula:FeCl₃·6H₂O
MDL Number:MFCD00149712
:270.3 g/mol
Merck Index:13,04048
Boiling Pt:280 - 285 °C (1013 hPa)
Melting Pt:37 °C
Density:1.82 g/cm³ (20 °C)
UN:3260
ADR:8,III
:M-Clarity™ quality level = MQ300
EINECS:231-729-4
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay (iodometric, FeCl₃·6H₂O):99.0 - 102.0 %
- Identity:passes test
- Insoluble matter:≤ 0.01 %
- Free chlorine:passes test
- Acidic substances:passes test
- Nitrate (NO₃):≤ 0.01 %
- Sulfate (SO₄):≤ 0.01 %
- Total nitrogen (N):≤ 0.001 %
- Total phosphorus (as PO₄):≤ 0.01 %
- Heavy metals (as Pb):≤ 0.005 %
- Ca (Calcium):≤ 0.01 %
- Cu (Copper):≤ 0.003 %
- Fe II (Iron II):≤ 0.002 % (The Fe(II)-content increases if the recommended storage conditions (+15 to +25 °C) are not observed)
- K (Potassium):≤ 0.005 %
- Mg (Magnesium):≤ 0.005 %
- Na (Sodium):≤ 0.05 %
- Zn (Zinc):≤ 0.003 %
Specification Test Results
Frequently Bought Together