Order Entry
United States
ContactUsLinkComponent
Zinc, Certipur® Reag. Ph. Eur. volumetric standard, secondary reference material, for complexometry traceable to NIST, Supelco®
Zinc, Certipur® Reag. Ph. Eur. volumetric standard, secondary reference material, for complexometry traceable to NIST, Supelco®
Catalog # EM1.02409.0100
Supplier:  MilliporeSigma
CAS Number:  
Zinc, Certipur® Reag. Ph. Eur. volumetric standard, secondary reference material, for complexometry traceable to NIST, Supelco®
Catalog # EM1.02409.0100
Supplier:  MilliporeSigma
Supplier Number:  1.02409.0100
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, pre-packed
  • Size:
    100 g
  • Cat. No.:
    EM1.02409.0100
  • Supplier no.:
    1.02409.0100

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    419 °C
  • Molar Mass:
    65.39 g/mol
  • Boiling Point:
    906 °C (1013 hPa)
  • Density:
    7.13 g/cm3 (20 °C)

Specification Test Results

About this item

 :
 :65.39 g/mol
Merck Index:12,10255
Boiling Pt:906 °C (1013 hPa)
Melting Pt:419 °C
Density:7.14 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-175-3
UN:3077
ADR:9,III
 :GHS09
CAS Number:7440-66-6
Formula:Zn
MDL Number:MFCD00011291