Calcein indicator for metal titration, Supelco®
Catalog # EM1.02315.0005
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Glass bottle, pre-packed
- Size:5 g
- Cat. No.:EM1.02315.0005
- Supplier no.:1.02315.0005
Specifications
Specification Test Results
- Solubility:(20 °C) slightly soluble
- Molar Mass:622.55 g/mol
- Bulk Density:600 kg/m3
Specification Test Results
About this item
:
:622.53 g/mol
EINECS:215-957-1
CAS Number:1461-15-0
:154071-48-4
Formula:C₃₀H₂₆N₂O₁₃
MDL Number:MFCD00005049