

Bismuth(III) nitrate basic, EMSURE® for analysis, Supelco®
Catalog # EM1.01878.0100
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:100 g
- Cat. No.:EM1.01878.0100
- Supplier no.:1.01878.0100
Specifications
Specification Test Results
- Solubility:(20 °C) practically insoluble
- Molar Mass:1461.99 g/mol
- Bulk Density:600 kg/m3
- Density:4.93 g/cm3 (20 °C)
- pH:3.2 (50 g/l, H2O, 20 °C) (slurry)
- Hazard Class:5.1
- Hazard Class Description:Oxidizer
Specification Test Results
About this item
:
:1461.99 g/mol
Density:4.928 g/cm³ (20 °C)
EINECS:215-136-8
UN:1477
ADR:5.1,II
:GHS03
:GHS07
CAS Number:1304-85-4
Formula:Bi₅O(OH)₉(NO₃)₄
MDL Number:MFCD00064844