
Barium fluoride, Suprapur® for trace metal analysis, Supelco®
Catalog # EM1.01722.0050
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:50 g
- Cat. No.:EM1.01722.0050
- Supplier no.:1.01722.0050
Specifications
Specification Test Results
- Solubility:1.2 g/l (25 °C)
- Melting Point:1353 °C
- Molar Mass:175.33 g/mol
- Boiling Point:2260 °C
- Density:4.89 g/cm3 (20 °C)
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials
Specification Test Results
About this item
:
Merck Index:12,01003
Boiling Pt:2260 °C (1013 hPa)
Melting Pt:1270 °C
Density:4.89 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-108-0
UN:1564
ADR:6.1,III
:175.33 g/mol
:GHS07
CAS Number:7787-32-8
Formula:BaF₂
MDL Number:MFCD00003450