Corning® Mouse Laminin
47743-734
: Corning
:
- Species:Mouse
- Size:1 mg
- Environmentally Preferable:Low Manufacturing Impact
- Environmental Certification:ISO 14001 - Bedford (reg. no. 14.4942)
- Protein/Peptide Name:Laminin
- Cat. No.:47743-734
- Supplier no.:354232
Laminin, a major component of basement membranes, has numerous biological activities including promotion of cell adhesion, migration, growth, and differentiation, including neurite outgrowth. It can be used as a thin coating on tissue culture surfaces or as a soluble additive to culture medium. The Laminin/Entactin Complex, high concentration (HC) is a special formulation that has been developed for three-dimensional (3D) culture.
Quality controlled by SDS-PAGE and tested and found negative for bacteria, fungi, and mycoplasma.