Flat Bottom Test Tubes, Globe Scientific
Supplier: Globe Scientific
Certificates
About this item
Ideal for use as a secondary tube for barcoding on automated analyzers.
- Flat bottom, self-standing tubes
- Available in clear polystyrene or unbreakable polypropylene
The 3 ml polystyrene tubes are graduated with a rim.
Popular brands of analyzers include Hitachi®, Olympus®, Corning®, Ortho®, Kodak®, Beckman®, IA®, Tosoh®, Vitros® and more.
Related Information
Globe Scientific Tubes Caps and Racks Laboratory Supplies Brochure
Product Literature
Frequently Bought Together