Inoculation Loops with Needle, Globe Scientific
Supplier: Globe Scientific
Certificates
About this item
Loops offer the most convenient and safest way of streaking culture media
- Polystyrene (PS)
- Certificate of calibration comes with each case of product
- Gamma sterilized
- Rigid feel for comfortable, stable, uniform streaking
Loops are sterilized by gamma radiation. Supplied with calibration certificate.
Molded from a special medical grade polystyrene, they offer a strong, rigid feel to provide comfortable, stable, and uniform streaking.
Frequently Bought Together