Snap Cap with Two Thumb Tabs, for 16 mm Tubes, Globe Scientific
Supplier: Globe Scientific
Certificates
About this item
An effective method to recap blood drawing tubes, glass test tubes, and plastic culture tubes
- Low density polyethylene (LDPE)
- Assorted colors available
- Fits both glass and plastic tubes
The snap caps are precision molded from low-density polyethylene (LDPE) and maintain a tight seal during centrifugation, agitation, and refrigeration procedures. Available in a variety of bright and attractive colors for sample identification purposes.
Ordering information: Snap caps are supplied in a pack of 1000.
Related Information
Globe Scientific Tubes Caps and Racks Laboratory Supplies Brochure
Product Literature
Frequently Bought Together