ContactUsLinkComponent
Protexidase™, Peroxidase Protective Buffer, Immuno™, MP Biomedicals
Protexidase™, Peroxidase Protective Buffer, Immuno™, MP Biomedicals
Catalog # IC980631
Supplier:  MP Biomedicals
CAS Number:  
Protexidase™, Peroxidase Protective Buffer, Immuno™, MP Biomedicals
Catalog # IC980631
Supplier:  MP Biomedicals
Supplier Number:  08980631
CAS Number:  
Specifications
  • Size:
    500 mL
  • Cat. No.:
    IC980631
  • Supplier no.:
    980631
Specifications
About this item
A peroxidase protective buffer which stabilizes HRP conjugates
It is compatible with all common blocking agents including Bovine Serum Albumin, skim milk powder, and Polysorbate 20. Protexidase provides stability of the HRP enzyme and it also inhibits micr
Protexidase buffer is 500 mL of PBS, pH 7.2, containing 0.5% BSA, antimicrobial preservatives and proprietary stabilizing compounds.