ImmunO™ Micro-Ouchterlony Plates, MP Biomedicals
Catalog # IC642771
Supplier: MP Biomedicals
Specifications
- No. of wells:56
- Well volume:10 μL
- Description:Micro-Ouchterlony Plates
- Cat. No.:IC642771
- Packaging:5 Plates
- Supplier no.:642771
Specifications
About this item
The MP ouchterlony plate is designed for specific but low affinity antisera
- Typically dilutions of 1:600 to 1:16000 are required
Each immunodiffusion plate contains 8 ouchterlony patterns consisting of a center well and 6 outer wells.The high quality immunodiffusion grade agarose contains 5% PEG which greatly enhances the sensitivity of the immunodiffusion assay. The plates contain 1.2% agarose (buffer - PBS with 0.1% sodium azide and 0.01% merthiolate).
Such antisera require properly spaced wells and polyethylene glycol in the agarose for optimal immuno-precipitation. The plates are prepared especially for the MP polyclonal antisera to human IgG subclasses.