Order Entry
United States
ContactUsLinkComponent
Trichoderma viride Cellulase Y-C, MP Biomedicals
Trichoderma viride Cellulase Y-C, MP Biomedicals
Catalog # IC320961
Supplier:  MP Biomedicals
CAS Number:  
Trichoderma viride Cellulase Y-C, MP Biomedicals
Catalog # IC320961
Supplier:  MP Biomedicals
Supplier Number:  08320961
CAS Number:  

Specifications

  • Species:
    Trichoderma viride
  • Size:
    10 g
  • Enzyme Name:
    Cellulase Y-C
  • Cat. No.:
    IC320961
  • Supplier no.:
    320961

Specifications

About this item

From Trichoderma viride This enzyme shows a very high filter paper decomposing activity, as well as, appreciable xylanase and hemicellulase activity

It is an effective enzyme for use with pectolyase Y-23 for plant cell wall removal.