ContactUsLinkComponent
THERMOPOL® Reaction Buffer Pack , New England Biolabs
THERMOPOL® Reaction Buffer Pack , New England Biolabs
Catalog # 101227-498
Small Business Enterprise
CAS Number:  
undefined
THERMOPOL® Reaction Buffer Pack , New England Biolabs
Catalog # 101227-498
Supplier Number:  B9004S
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    ThermoPol Reaction Buffer Pack
  • Size:
    6.0 mL
  • Cat. No.:
    101227-498
  • Supplier no.:
    B9004S
Specifications
About this item
10X ThermoPol Reaction Buffer is optimized for use with VentR® and Deep VentR™ DNA Polymerases
This buffer also provides superior reaction conditions for other thermophilic DNA polymerases, including Taq DNA Polymerase, as well as various other DNA and RNA modifying enzymes. Supplemental MgSO4 is provided to maximize flexibility for users that require additional Mg2+ for their specific application.