ContactUsLinkComponent
Taq 5X Master Mix, New England Biolabs
Taq 5X Master Mix, New England Biolabs
Catalog # 101444-748
Small Business Enterprise
CAS Number:  
undefined
Taq 5X Master Mix, New England Biolabs
Catalog # 101444-748
Supplier Number:  M0285L
Small Business Enterprise
CAS Number:  
Specifications
  • Description:
    Taq 5X Master Mix
  • Size:
    500 Reactions
  • Cat. No.:
    101444-748
  • Supplier no.:
    M0285L
Specifications
About this item
Taq 5X Master Mix is an optimized ready-to-use solution containing Taq DNA Polymerase, dNTPs, MgCl2, KCl and stabilizers
It is best for routine PCR applications from templates including pure DNA solutions, bacterial colonies, and cDNA. It can amplify up to 4 kb from complex genomic DNA or up to 5 kb from lambda DNA.