- Description:SNAP-Surface Block
- Size:200 nmol
- Cat. No.:101447-698
- Supplier no.:S9143S
SNAP-Surface® Block (C8 propanoic acid benzylguanine, CBG) is a nonfluorescent compound that blocks the reactivity of the SNAP-tag® in solution or on the surface of living cells
It can be used to generate inactive controls in live cell labeling experiments performed with SNAP-tag fusion proteins. SNAP-Surface Block reacts with the SNAP-tag irreversibly, inactivating it for subsequent labeling steps. This blocker is largely membrane impermeable essentially limiting blocking to cell surface-exposed SNAP-tags.