Specifications
- Description:SNAP-Surface Alexa Fluor 546
- Size:50 nmol
- Cat. No.:101447-680
- Supplier no.:S9132S
Specifications
About this item
SNAP-Surface Alexa Fluor 546 is a photostable fluorescent substrate that can be used to label SNAP-tag® fusion proteins in solution or on the surface of living cells
This cell impermeable substrate is based on Alexa Fluor 546 and is suitable for standard TAMRA or Cy3 filter sets. It has an excitation maximum at 558 nm and emission maximum at 574 nm. This package includes 50 nmol of SNAP-Surface Alexa Fluor 546 substrate, sufficient to make 10 ml of a 5 µM SNAP-tag fusion protein labeling solution.