Order Entry
United States
ContactUsLinkComponent
ScaI-HF®, Restriction Enzymes, New England Biolabs
ScaI-HF®, Restriction Enzymes, New England Biolabs
Catalog # 101641-118
Small Business Enterprise
CAS Number:  
undefined
ScaI-HF®, Restriction Enzymes, New England Biolabs
Catalog # 101641-118
Supplier Number:  R3122M
Small Business Enterprise
CAS Number:  

Specifications

  • Concentration:
    100000 U/ml
  • Description:
    ScaI-HF®
  • Size:
    5000 units
  • Enzyme Name:
    ScaI-HF
  • Cat. No.:
    101641-118
  • Supplier no.:
    R3122M

Specifications

About this item

An E.coli strain that carries the cloned and modified ScaI gene from Streptomyces caespitosus (H. Takahashi).

High Fidelity (HF®) Restriction Enzymes have 100% activity in CutSmart® Buffer; single-buffer simplicity means more straightforward and streamlined sample processing. HF enzymes also exhibit dramatically reduced star activity. HF enzymes are all Time-Saver™ qualified and can therefore cut substrate DNA in 5-15 with the flexibility to digest overnight without degradation to DNA. Engineered with performance in mind, HF restriction enzymes are fully active under a broader range of conditions, minimizing off-target products, while offering flexibility in experimental design.