PUREXPRESS® Disulfide Bond Enhancer, New England Biolabs
Catalog # 101640-914
Supplier: New England Biolabs (NEB)
Small Business Enterprise
CAS Number:
Specifications
- Description:PURExpress Disulfide Bond Enhancer
- Size:50 Reactions
- Cat. No.:101640-914
- Supplier no.:E6820S
Specifications
About this item
The PURExpress® Disulfide Bond Enhancer (PDBE) is a proprietary blend of proteins and buffer components designed to correctly fold target proteins with multiple disulfide bonds produced in PURExpress reactions or E.coli S30 extracts.
Added at the beginning of a reaction, the components promote a proper disulfide bond pattern by assisting with the oxidation of cysteine thiols and correcting mis-oxidized substrates. These enhancements can increase the yield of soluble and functionally active protein.