About this item
The Phusion High-Fidelity PCR Kit (large) contains a sufficient supply of Phusion High-Fidelity DNA Polymerase, Phusion HF and GC Buffers, deoxynucleotides, magnesium chloride, DMSO, and DNA size standard to perform 200 reactions
Control template and primers are provided for 20 control reactions.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Application:
- Environmentally Preferable: