Orders LinkContactUsLinkComponent
Nt.AlwI
Nt.AlwI
Nt.AlwI, 500 units
Catalog # 101229-484
Small Business Enterprise
Supplier #:  R0627S
Nt.AlwI
Catalog # 101229-484
Small Business Enterprise
CAS Number:  
Nt.AlwI
Nt.AlwI
Nt.AlwI, 500 units
CAS Number:  
Price
$85.00
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
An E.coli strain containing a chimeric gene encoding the DNA recognition domain of AlwI and the cleavage/dimerization domain of Nt.BstNBI.
Specifications
Specifications
  • Description:
    Nt.AlwI
  • Size:
    500 units
  • Cat. No.:
    101229-484
  • Enzyme Name:
    Nt.AlwI