Order Entry
United States
ContactUsLinkComponent
NEBNext® Magnesium RNA Fragmentation Module, New England Biolabs
NEBNext® Magnesium RNA Fragmentation Module, New England Biolabs
Catalog # 101710-296
Small Business Enterprise
CAS Number:  
undefined
NEBNext® Magnesium RNA Fragmentation Module, New England Biolabs
Catalog # 101710-296
Supplier Number:  E6150S
Small Business Enterprise
CAS Number:  

Specifications

  • Description:
    NEBNEXT® Magnesium RNA Fragmentation Module
  • Cat. No.:
    101710-296
  • No. of reactions:
    200
  • Supplier no.:
    E6150S

Specifications

About this item

The NEBNext® Magnesium RNA Fragmentation Module has been optimized to fragment RNA into small pieces using divalent cations under elevated temperature

The NEBNext Magnesium RNA Fragmentation Module contains buffers that are suited for RNA fragmentation. Each of these components must pass rigorous quality control standards.